Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 mL | Check Cart for Availability | € 878,00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
packaging
pkg of 1 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecules)
format
neat
storage temp.
−20°C
SMILES string
N(N=O)(CCCC(=O)O)C
InChI
1S/C5H10N2O3/c1-7(6-10)4-2-3-5(8)9/h2-4H2,1H3,(H,8,9)
InChI key
SJLBIPLIGYWGJV-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including certificate/ product information sheets, please go to the website of the issuing Pharmacopoeia.
Application
It is also used to prepare standard, nitrosamine standards stock solution mixture, and sensitivity solutions to determine NMBA impurity in drug substances and drug products (valsartan, irbesartan, and losartan potassium etc) by chromatography according to general chapter <1469> of United States Pharmacopeia.
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| application(s) pharmaceutical (small molecules) | application(s) pharmaceutical | application(s) pharmaceutical (small molecules) | application(s) USP Biologics |
| format neat | format - | format neat | format single component solution (solution in methanol) |
| grade pharmaceutical primary standard ( | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard ( | grade pharmaceutical primary standard ( |
| storage temp. −20°C | storage temp. - | storage temp. −20°C | storage temp. −20°C |
| manufacturer/tradename USP | manufacturer/tradename Manufactured by: Sigma-Aldrich Production GmbH, Switzerland | manufacturer/tradename USP | manufacturer/tradename USP |
| packaging pkg of 1 mg | packaging pkg of 1 mL | packaging pkg of 1 mg | packaging pkg of 1 mg |
Still not finding the right product?
Explore all of our products under N-Nitrosomethylaminobutyric Acid (NMBA)
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
35.6 °F
flash_point_c
2 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Eine Übersicht über Prüfungen auf Nitrosaminverunreinigungen, weltweite Vorschriften und wichtige Überlegungen bei der Filterauswahl für die Probenvorbereitung und -analyse.
This application note describes the LC-MS-based quantitative analysis of known nitrosamine impurities following procedures 1 and 3 as given in USP general chapter <1469>.
An overview of nitrosamine impurity testing, worldwide regulations, and key considerations in filter selection for sample preparation and analysis.
Related Content
Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.
Bestellen Sie aus einer breiten Palette hochgradig charakterisierter Materialien als primäre Referenzstandards zur Verwendung mit USP-NF-Monographien für die Prüfung von Arzneistoffen und Darreichungsformen, pharmazeutischen Hilfsstoffen, Lebensmittelinhaltsstoffen und Nahrungsergänzungsmitteln.
Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1466696-1ML | 04065265949798 |




