Skip to Content
Merck

Skip To

1393005

USP

Mesoridazine besylate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Mesoridazine benzenesulfonate, 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylsulfinyl)-10H-phenothiazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 408,00

About This Item

Empirical Formula (Hill Notation):
C27H32N2O4S3
CAS Number:
Molecular Weight:
544.75
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 408,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

pharmaceutical primary standard

API family

mesoridazine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OS(=O)(=O)c1ccccc1.CN2CCCCC2CCN3c4ccccc4Sc5ccc(cc35)S(C)=O

InChI

1S/C21H26N2OS2.C6H6O3S/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(26(2)24)15-19(21)23;7-10(8,9)6-4-2-1-3-5-6/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3;1-5H,(H,7,8,9)

InChI key

CRJHBCPQHRVYBS-UHFFFAOYSA-N

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
166250414421001663008
format

neat

format

neat

format

neat

format

neat

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

API family

mesoridazine

API family

thioridazine

API family

metronidazole

API family

thioridazine

Gene Information

human ... DRD2(1813), HTR2A(3356)

Gene Information

-

Gene Information

-

Gene Information

human ... DRD2(1813), DRD3(1814), DRD4(1815), HTR2A(3356), HTR2C(3358)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mesoridazine besylate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Mesoridazine benzenesulfonate is a D2 dopamine receptor antagonist; antipsychotic.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
1393005-25MG04065265963237
1393005-250MG04061838732798

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service