Skip to Content
Merck

Skip To

1369055

USP

Lithol Rubine BK

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-Hydroxy-4-[(4-methyl-2-sulfophenyl)azo]-2-naphthoic acid calcium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H12CaN2O6S
CAS Number:
Molecular Weight:
424.44
UNSPSC Code:
41116107
NACRES:
NA.24
Colour Index Number:
15850
Product 1369055 is currently not available for purchase. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

lithol rubine

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

SMILES string

[Ca++].Cc1ccc(N=Nc2c(O)c(cc3ccccc23)C([O-])=O)c(c1)S([O-])(=O)=O

InChI

1S/C18H14N2O6S.Ca/c1-10-6-7-14(15(8-10)27(24,25)26)19-20-16-12-5-3-2-4-11(12)9-13(17(16)21)18(22)23;/h2-9,19H,1H3,(H,22,23)(H,24,25,26);/q;+2/p-2/b20-16-;

InChI key

WANIKCLUTOABTP-LRZQPWDCSA-L

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lithol Rubine BK USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1048277906891048266
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

analytical standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

pharmaceutical (small molecule)

API family

lithol rubine

API family

balsalazide

API family

-

API family

balsalazide


Still not finding the right product?

Explore all of our products under Lithol Rubine BK


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1369055-100MG04061833020210

Questions

Reviews

No rating value

Active Filters