Skip to Content
Merck

Skip To

1350104

USP

Isopropyl acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Acetic acid isopropyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
3 x 1.2 mL
Check Cart for Availability
€ 399,00

About This Item

Linear Formula:
CH3COOCH(CH3)2
CAS Number:
Molecular Weight:
102.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1740761

€ 399,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

3.5 (vs air)

vapor pressure

47 mmHg ( 20 °C)

API family

isopropyl acetate

autoignition temp.

894 °F

expl. lim.

1.8 %, 37 °F, 8 %

manufacturer/tradename

USP

refractive index

n20/D 1.377 (lit.)

bp

85-91 °C (lit.)

mp

−73 °C (lit.)

density

0.872 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)OC(C)=O

InChI

1S/C5H10O2/c1-4(2)7-5(3)6/h4H,1-3H3

InChI key

JMMWKPVZQRWMSS-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isopropyl acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
900519537462112992
grade

pharmaceutical primary standard

grade

ZerO2®

grade

-

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

-

format

neat

format

-

format

-

format

-

bp

85-91 °C (lit.)

bp

85-91 °C (lit.)

bp

85-91 °C (lit.)

bp

85-91 °C (lit.)

expl. lim.

1.8 %, 37 °F

expl. lim.

1.8 %, 37 °F

expl. lim.

1.8 %, 37 °F

expl. lim.

1.8 %, 37 °F


Still not finding the right product?

Explore all of our products under Isopropyl acetate


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

41.0 °F - closed cup

flash_point_c

5 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters