Skip to Content
Merck

Skip To

1325005

USP

Hydroxyamphetamine hydrobromide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-(2-Aminopropyl)phenol hydrobromide, NSC 61065, Paredrine hydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 426,00

About This Item

Empirical Formula (Hill Notation):
C9H13NO · HBr
CAS Number:
Molecular Weight:
232.12
MDL number:
UNSPSC Code:
41116107
EC Number:
206-181-4
NACRES:
NA.24

€ 426,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

hydroxyamphetamine

manufacturer/tradename

USP

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Br-].[N+H3]C(Cc1ccc(cc1)O)C

InChI

1S/C9H13NO.BrH/c1-7(10)6-8-2-4-9(11)5-3-8;/h2-5,7,11H,6,10H2,1H3;1H

InChI key

RZCJLMTXBMNRAD-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydroxyamphetamine hydrobromide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
10788461610001BP199
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

BP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

format

neat

format

neat

format

neat

API family

hydroxyamphetamine

API family

bupropion

API family

scopolamine, hyoscine

API family

hyoscine, scopolamine

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

drug control

-

drug control

-

drug control

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1325005-200MG04061833831717

Questions

Reviews

No rating value

Active Filters