Skip to Content
Merck

Skip To

1296031

USP

Glycopyrrolate Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H25NO3
CAS Number:
Molecular Weight:
303.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

glycopyrrolate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N1(CC(CC1)OC(=O)C(O)(C3CCCC3)c2ccccc2)C

InChI

1S/C18H25NO3/c1-19-12-11-16(13-19)22-17(20)18(21,15-9-5-6-10-15)14-7-3-2-4-8-14/h2-4,7-8,15-16,21H,5-6,9-13H2,1H3

InChI key

OVGMKPGXRHJNKJ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glycopyrrolate Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Glycopyrrolate
  • Glycopyrrolate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2468PHR2466PHR2465
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical

application(s)

pharmaceutical

format

neat

format

-

format

-

format

-

API family

glycopyrrolate

API family

glycopyrrolate

API family

glycopyrrolate

API family

glycopyrrolate

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-30°C


Still not finding the right product?

Explore all of our products under Glycopyrrolate Related Compound B


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1296031-50MG04061835346059

Questions

Reviews

No rating value

Active Filters