Skip to Content
Merck

Skip To

1269243

USP

Famotidine Related Compound D

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-{{{2-[(Diaminomethylene)amino]thiazol-4-yl}methyl}sulphanyl}propanamide, 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 1.400,00

About This Item

Empirical Formula (Hill Notation):
C8H13N5OS2
CAS Number:
Molecular Weight:
259.35
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 1.400,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

grade

pharmaceutical primary standard

agency

USP

API family

famotidine

form

powder

packaging

pkg of 25 mg

manufacturer/tradename

USP

storage condition

protect from light

color

white

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)CSCCC(=O)N)N=CNN

InChI

1S/C8H13N5OS2/c9-7(14)1-2-15-3-6-4-16-8(13-6)11-5-12-10/h4-5H,1-3,10H2,(H2,9,14)(H,11,12,13)

InChI key

ASHSLSDRNNWQKI-UHFFFAOYSA-N

General description

Famotidine Related Compound D (Famotidine Impurity D) is an impurity of famotidine, which is a hydrophilic, cationic, histamine H2 receptor antagonist drug that effectively inhibits gastric acid secretion in humans.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Famotidine Related Compound D USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Famotidine

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1269265PHR29111269232
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

form

powder

form

powder

form

-

form

powder

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

storage condition

protect from light

storage condition

protect from light

storage condition

-

storage condition

protect from light


Still not finding the right product?

Explore all of our products under Famotidine Related Compound D


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Stability indicating method for famotidine in pharmaceuticals using porous graphitic carbon column
Helali N and Monser L
Journal of Separation Science, 31(2) (2008)
Famotidine
Pharmacopeia, US
United States Pharmacopeia, 39(1)(1-2), 1815-1815 (2020)
Capillary zone electrophoresis method for the determination of famotidine and related impurities in pharmaceuticals
Helali N, et al.
Talanta, 74(4) (2008)



Global Trade Item Number

SKUGTIN
PH004000-1MG04061822877665
1269243-25MG04061833830239

Questions

Reviews

No rating value

Active Filters