Skip to Content
Merck

Skip To

1268604

USP

Etidronic acid monohydrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1-Hydroxyethylidene)bisphosphonic acid monohydrate, 1-Hydroxyethylidenediphosphonic acid

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C2H8O7P2 · H2O
CAS Number:
Molecular Weight:
224.04
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:


grade

pharmaceutical primary standard

API family

etidronic acid

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.CC(O)(P(O)(O)=O)P(O)(O)=O

InChI

1S/C2H8O7P2.H2O/c1-2(3,10(4,5)6)11(7,8)9;/h3H,1H3,(H2,4,5,6)(H2,7,8,9);1H2

InChI key

KKNZXUHBWLPUFN-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etidronic acid monohydrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Etidronate Disodium
  • Etidronate Disodium Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0000572543421233508
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

-

format

neat

API family

etidronic acid

API family

etidronate

API family

-

API family

edetic acid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under Etidronic acid monohydrate


pictograms

CorrosionEnvironment

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1268604-1G04061833859896

Questions

Reviews

No rating value

Active Filters