Skip to Content
Merck

Skip To

1265752

USP

Ethyl laurate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ethyl dodecanoate, Ethyl laurate, Lauric acid ethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 mg
Check Cart for Availability
€ 247,00

About This Item

Linear Formula:
CH3(CH2)10COOC2H5
CAS Number:
Molecular Weight:
228.37
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1769671

€ 247,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

ethyl laurate

manufacturer/tradename

USP

bp

269 °C (lit.)

density

0.862 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCC(=O)OCC

InChI

1S/C14H28O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h3-13H2,1-2H3

InChI key

MMXKVMNBHPAILY-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethyl laurate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W244104W24411207523
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

FG, Fragrance grade, Halal, Kosher

grade

FG, Kosher, natural

grade

analytical standard

format

neat

format

-

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

flavors and fragrances

application(s)

flavors and fragrances

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

density

0.862 g/mL at 20 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

density

0.862 g/mL at 20 °C (lit.)

bp

269 °C (lit.)

bp

269 °C (lit.)

bp

269 °C (lit.)

bp

269 °C (lit.)


Still not finding the right product?

Explore all of our products under Ethyl laurate


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1265752-500MG04061833829974

Questions

Reviews

No rating value

Active Filters