Skip to Content
Merck

Skip To

1265402

USP

Ethyl acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

EtOAc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
3 x 1.2 mL
Check Cart for Availability
€ 399,00

About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
506104

€ 399,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

manufacturer/tradename

USP

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

General description

Ethyl acetate, a volatile ester, is frequently employed in microbiology as a solvent and extraction agent. In the pharmaceutical and biologics industries, it is particularly useful in processes where a non-polar solvent is needed to extract bioactive compounds or for purification steps. Ethyl acetate is valued for its relatively low toxicity and efficient solvent properties, contributing to the formulation of biologic products while maintaining quality and compliance with regulatory standards.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethyl acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pha
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR14811.00864EX0240
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

ACS reagent, reagent grade

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

liquid formulation
pharmaceutical
semi-solid formulation

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

format

neat

format

neat

format

neat

form

liquid

form

-

form

liquid

form

liquid

refractive index

n20/D 1.3720 (lit.)

refractive index

n20/D 1.3720 (lit.)

refractive index

n20/D 1.3720 (lit.)

refractive index

n20/D 1.3720 (lit.)


Still not finding the right product?

Explore all of our products under Ethyl acetate


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters