Skip to Content
Merck

Skip To

1248218

USP

Erythropoietin

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C809H1301N229O240S5
CAS Number:
Molecular Weight:
18235.70
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

erythropoietin

form

solid

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical

General description

Erythropoietin (EPO) is a glycoprotein hormone produced by the kidneys to regulate red blood cell production. Recombinant EPO is used in treating anemia due to chronic kidney disease or chemotherapy. The USP standard supports bioidentity and potency testing using validated cell-based assays.

The USP biologics proteins category provides critical standards and reference materials necessary for the development and manufacturing of therapeutic proteins. These proteins, which can be naturally derived or genetically engineered, are essential in treating various diseases and conditions, ensuring that they meet quality, safety, and efficacy standards.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Erythropoietin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Epoetin

Other Notes

This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E11000001248200Y0002009
application(s)

USP Biologics
pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

-

form

solid

form

solid

form

solid

form

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

USP

manufacturer/tradename

EDQM

agency

USP

agency

EP

agency

-

agency

-

API family

erythropoietin

API family

ergosterol

API family

erythropoietin

API family

erythropoietin


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Bestellen Sie aus einer breiten Palette hochgradig charakterisierter Materialien als primäre Referenzstandards zur Verwendung mit USP-NF-Monographien für die Prüfung von Arzneistoffen und Darreichungsformen, pharmazeutischen Hilfsstoffen, Lebensmittelinhaltsstoffen und Nahrungsergänzungsmitteln.

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.


Epoetin
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(5), 1657-1657 (2018)



Global Trade Item Number

SKUGTIN
1248218-0.1MG04065265966337
1248218-100UG04065272601016

Questions

Reviews

No rating value

Active Filters