Skip to Content
Merck

Skip To

1235933

USP

Entacapone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Cyano-N,N-diethyl-3-(3,4-dihydroxy-5-nitrophenyl)propenamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 777,00

About This Item

Empirical Formula (Hill Notation):
C14H15N3O5
CAS Number:
Molecular Weight:
305.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 777,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

entacapone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[N+](=O)([O-])c1c(c(cc(c1)\C=C(\C(=O)N(CC)CC)/C#N)O)O

InChI

1S/C14H15N3O5/c1-3-16(4-2)14(20)10(8-15)5-9-6-11(17(21)22)13(19)12(18)7-9/h5-7,18-19H,3-4H2,1-2H3/b10-5+

InChI key

JRURYQJSLYLRLN-BJMVGYQFSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Entacapone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Entacapone Tablets

Biochem/physiol Actions

Entacapone is a catechol-O-methyl transferase (COMT) inhibitor. Used in treatment of Parkinson′s disease, entacapone is administered with L-DOPA to inihibit COMT from converting L-DOPA into a compound that cannot cross the blood brain barrier.
Entacapone is a catechol-O-methyl transferase (COMT) inhibitor. Antiparkinson; anti-Dyskinesia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y00012491235944SML0654
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

USP

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

API family

entacapone

API family

entacapone

API family

entacapone

API family

-


Still not finding the right product?

Explore all of our products under Entacapone


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters