Skip to Content
Merck

Skip To

1140101

USP

Clomiphene Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(E/Z)-2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethylethanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H29NO · HCl
CAS Number:
Molecular Weight:
407.98
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

clomifene

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.N(CCOc1ccc(cc1)\C(=C/c3ccccc3)\c2ccccc2)(CC)CC

InChI

1S/C26H29NO.ClH/c1-3-27(4-2)19-20-28-25-17-15-24(16-18-25)26(23-13-9-6-10-14-23)21-22-11-7-5-8-12-22;/h5-18,21H,3-4,19-20H2,1-2H3;1H/b26-21-;

InChI key

VPQKHXADDASFHZ-AURQPEIRSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clomiphene Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Clomiphene Citrate
  • Clomiphene Citrate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR374215344131140000
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

neat

format

neat

API family

clomifene

API family

-

API family

phenyltoloxamine

API family

clomifene


Still not finding the right product?

Explore all of our products under Clomiphene Related Compound A


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - STOT RE 2 Oral

target_organs

ovary

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1140101-100MG04061833842607
1140101-25MG04065270799722

Questions

Reviews

No rating value

Active Filters