Skip to Content
Merck

Skip To

1098093

USP

Ceftiofur System Suitability Mixture

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 1.470,00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

€ 1.470,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

ceftiofur

manufacturer/tradename

USP

application(s)

pharmaceutical

format

mixture

storage temp.

−20°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ceftiofur System Suitability Mixture USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ceftiofur Hydrochloride
  • Ceftiofur Sodium

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
103324712705261096994
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

format

mixture

format

mixture

format

mixture

format

mixture

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

API family

ceftiofur

API family

ampicillin

API family

fingolimod

API family

cefadroxil

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
109809304061833849392
1098093-50MG04061838669520

Questions

Reviews

No rating value

Active Filters