Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 mL | Check Cart for Availability | € 756,00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
cannabis
manufacturer/tradename
USP
concentration
1 mL (in acetonitrile and triethylamine with stabilizer)
application(s)
pharmaceutical
General description
Application
Other Notes
- Cannabidiolic acid [CAS 1244-58-2]
- Tetrahydrocannabinolic acid [CAS 23978-85-0]
- Cannabidivarinic acid [CAS 31932-13-5]
- Cannabigerolic acid [CAS 25555-57-1]
- Tetrahydrocannabivarinic acid [CAS 39986-26-0]
1 of 1
This Item | |||
|---|---|---|---|
| application(s) pharmaceutical | application(s) - | application(s) pharmaceutical (small molecule) | application(s) forensics and toxicology |
| concentration 1 mL (in acetonitrile and triethylamine with stabilizer) | concentration 1 mL in methanol | concentration 1.0 mg/mL in methanol | concentration - |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - |
| API family cannabis | API family cannabis | API family - | API family - |
microbialanalysis | microbialanalysis | microbialanalysis | microbialanalysis |
signalword
Danger
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
14.0 °F
flash_point_c
-10 °C
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1089172-1ML | 04065265966290 |





