Skip to Content
Merck

Skip To

1087472

USP

Calycosin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3′,7-Dihydroxy-4′-methoxy-isoflavone, 3′-Hydroxyformononetin, Cyclosin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 mg
Check Cart for Availability
€ 299,00

About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 299,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

calycosin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C(C(C=C2)=CC(O)=C2OC)=COC3=CC(O)=CC=C13

InChI

1S/C16H12O5/c1-20-14-5-2-9(6-13(14)19)15-8-12(18)11-4-3-10(17)7-16(11)21-15/h2-8,17,19H,1H3

InChI key

QAGGICSUEVNSGH-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Calycosin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Astragalus Root
  • Astragalus Root Dry Extract
  • Astragalus Root Powder

Biochem/physiol Actions

Isoflavone and phytoestrogen isolated froma traditional Chinese medicinal herb. Calycosin induces angiogenesis in HUVEC and promotes proliferation of estrogen-receptor positive cells in vivo and in vitro. Also functions as a vasorelaxant on rat thoracic aorta.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Calycosin


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1087472-50MG04061833841044

Questions

  1. 본 제품의 purity를 문의드립니다.

    1 answer
    1. This is a product of the United States Pharmacopeia. The current lot is reported to be 0.987 mg of calycosin per mg of material or 98.7% pure.

      Helpful?

Reviews

No rating value

Active Filters