Skip to Content
Merck

Skip To

1076352

USP

Bretylium tosylate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(o-Bromobenzyl)ethyldimethylammonium p-toluenesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 446,00

About This Item

Empirical Formula (Hill Notation):
C18H24BrNO3S
CAS Number:
Molecular Weight:
414.36
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 446,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

bretylium

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.CC[N+](C)(C)Cc2ccccc2Br

InChI

1S/C11H17BrN.C7H8O3S/c1-4-13(2,3)9-10-7-5-6-8-11(10)12;1-6-2-4-7(5-3-6)11(8,9)10/h5-8H,4,9H2,1-3H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

InChI key

KVWNWTZZBKCOPM-UHFFFAOYSA-M

Gene Information

human ... SLC6A2(6530)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bretylium tosylate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Bretylium Tosylate Injection
  • Bretylium Tosylate in Dextrose Injection

Biochem/physiol Actions

Bretylium tosylate has a biphasic response, initially inducing norepinephrine release followed by sympathetic ganglionic blockade; class III antiarrhythmic agent; competitive inhibitor of acetylcholinesterase.
Class III antiarrhythmic agent; competitive inhibitor of acetylcholinesterase.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
106100510121341076501
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

Gene Information

human ... SLC6A2(6530)

Gene Information

human ... CHRM1(1128)

Gene Information

-

Gene Information

-

API family

bretylium

API family

benztropine

API family

ademetionine

API family

bromocriptine


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters