Skip to Content
Merck

Skip To

1076013

USP

Bivalirudin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

D-Phenylalanyl-L-prolyl-L-arginyl-L-prolyl-glycyl-glycyl-glycyl-glycyl-L-asparagyl-glycyl-L-α-aspartylL-phenylalanyl-L-α-glutamyl-L-α-glutamyl-L-isoleucyl-L-prolyl-L-α-glutamyl-L-α-glutamyl-L-tyrosyl-L -leucine trifluoroacetate (salt) hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C98H138N24O33
Molecular Weight:
2180.29
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


API family

bivalirudin

form

solid

packaging

pkg of 5 mg

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

General description

Peptide family: Bivalirudin

A direct thrombin inhibitor used in anticoagulation therapy.

The USP biologics peptides category encompasses a diverse range of therapeutic peptides that are essential in managing various medical conditions. These peptides, typically consisting of amino acid sequences of 40 residues or less, are critical for the development of high-quality medicines. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of peptide therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Bivalirudin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Bivalirudin for Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
107603512385121238578
application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

form

solid

form

solid

form

solid

form

solid

format

neat

format

neat

format

neat

format

neat

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

packaging

pkg of 5 mg

packaging

pkg of 1 mg

packaging

pkg of 10 mg

packaging

pkg of 10 mg


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1076013-5MG04065266773880

Questions

Reviews

No rating value

Active Filters