Skip to Content
Merck

Skip To

1045993

USP

Azathioprine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Methyl-4-nitro-1H-imidazol-5-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
15 mg
Check Cart for Availability
€ 1.270,00

About This Item

Empirical Formula (Hill Notation):
C4H6N4O2
CAS Number:
Molecular Weight:
142.12
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 1.270,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

azathioprine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1nc[n](c1N)C

InChI

1S/C4H6N4O2/c1-7-2-6-4(3(7)5)8(9)10/h2H,5H2,1H3

InChI key

GJVMTMXQJDQJSS-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Azathioprine Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Azathioprine
  • Azathioprine Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
146600713383241643554
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

azathioprine

API family

nitrofurazone

API family

imiquimod

API family

temozolomide

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Azathioprine Related Compound A


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1045993-15MG04061833840665

Questions

Reviews

No rating value

Active Filters