Skip to Content
Merck

Skip To

1031401

USP

Amoxapine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Chloro-11-(1-piperazinyl)dibenz[b,f][1,4]oxazepine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 407,00

About This Item

Empirical Formula (Hill Notation):
C17H16ClN3O
CAS Number:
Molecular Weight:
313.78
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 407,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

amoxapine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1ccc2Oc3ccccc3N=C(N4CCNCC4)c2c1

InChI

1S/C17H16ClN3O/c18-12-5-6-15-13(11-12)17(21-9-7-19-8-10-21)20-14-3-1-2-4-16(14)22-15/h1-6,11,19H,7-10H2

InChI key

QWGDMFLQWFTERH-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amoxapine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Amoxapine Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1841A129A-123
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

certified reference material

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Cerilliant®

format

neat

format

neat

format

-

format

single component solution

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

-

application(s)

clinical testing

API family

amoxapine

API family

amoxapine

API family

-

API family

-

Gene Information

human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816), SLC6A2(6530), SLC6A4(6532)

Gene Information

human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816), SLC6A2(6530), SLC6A4(6532)

Gene Information

human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816), HTR2A(3356), HTR2B(3357), HTR2C(3358), SLC6A2(6530), SLC6A4(6532)

Gene Information

human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816), SLC6A2(6530), SLC6A4(6532)


Still not finding the right product?

Explore all of our products under Amoxapine


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1031401-200MG04061833839553

Questions

Reviews

No rating value

Active Filters