Skip to Content
Merck
All Photos(1)

Key Documents

44897-U

Supelco

ASTM® D6584 Tricaprin Solution

greener alternative

8000 μg/mL in pyridine, analytical standard

Synonym(s):

Biodiesel impurity

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Agency

ASTM® D6584

form

liquid

packaging

pkg of 1 × 5 mL

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

8000 μg/mL in pyridine

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

single component solution

greener alternative category

storage temp.

2-8°C

InChI

1S/C33H62O6/c1-4-7-10-13-16-19-22-25-31(34)37-28-30(39-33(36)27-24-21-18-15-12-9-6-3)29-38-32(35)26-23-20-17-14-11-8-5-2/h30H,4-29H2,1-3H3

InChI key

LADGBHLMCUINGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisabetta Esposito et al.
Pharmaceutical research, 25(7), 1521-1530 (2008-01-04)
The present investigation describes a formulative study for the development of innovative drug delivery systems for bromocriptine. Solid lipid nanoparticles (SLN) based on different lipidic components have been produced and characterized. Morphology and dimensional distribution have been investigated by electron
Matthew J Schaefer et al.
Biomaterials, 23(16), 3465-3471 (2002-07-09)
The purpose of this article is to examine the effects of tricaprin on the physical characteristics and in vitro release of etoposide from poly (lactic-co-glycolic acid) microspheres. The microspheres were synthesized through the use of a single-emulsion solvent-extraction procedure. Samples
Doron J Aframian et al.
Quintessence international (Berlin, Germany : 1985), 41(3), e36-e42 (2010-03-10)
Salivary gland impairment is a major problem that can result in hyposalivation and a decrease in quality of life. Causes for mouth dryness can be grossly classified into three major groups: iatrogenic, immunogenic, and metabolic. At present, insufficient therapies exist
Soo-Jeong Lim et al.
International journal of pharmaceutics, 243(1-2), 135-146 (2002-08-15)
Solid lipid nanoparticles (SLNs) have gained attention as a colloidal drug carrier, particularly for drugs with limited solubility. The poor aqueous solubility of all-trans retinoic acid (ATRA) has been a limiting factor in its clinical use. This study was undertaken
H Sato
The Journal of veterinary medical science, 56(4), 781-782 (1994-08-01)
Milk fortified with medium chain triglycerides (MCT) was given to neonatal calves to increase their energy intake. Tricaprylin (C8), tricaprin (C10), a mixture of C8 and C10 (mixed MCT) or soya oil were given at 11-18 days of age. Feeding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service