Skip to Content
Merck
All Photos(1)

Key Documents

CRM44918

Supelco

ASTM® D6584 Standard Solution Kit

with Internal Standards, certified reference material

Synonym(s):

TM=["ASTM"] D6584 reference standard set

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€588.00

€588.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€588.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€588.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

ASTM® D6584

product line

TraceCERT®

CofA

current certificate can be downloaded

concentration

in pyridine (varied concentration)

technique(s)

gas chromatography (GC): suitable

format

multi-component solution

storage temp.

2-8°C

Application

<ul>
<li><strong>Quantitative analysis of glycerol in biodiesel:</strong> The ASTM D6584 Standard Solution Kit can be used in the chromatographic procedure that was carried out an eco-friendly sonoluminescent method to determine free glycerol content in biodiesel samples (Diniz et al., 2013).</li>
</ul>

Other Notes

Our kit contains seven certified reference materials (CRMs) that are produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of these CRMs can be found on the certificate of analyses.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Kit Components Also Available Separately

Product No.
Description
SDS

  • ASTM® D6584 Standard solution 1, certified reference material, in pyridine (varied conc.)

  • CRM44914ASTM® D6584 Standard Solution 2, in pyridine (varied), ampule of 1 mLSDS

  • CRM44915ASTM® D6584 Standard Solution 3, certified reference material, in pyridine (varied), ampule of 1 mLSDS

  • CRM44916ASTM® D6584 Standard Solution 4, certified reference material, in pyridine (varied conc.)SDS

  • CRM44917ASTM® D6584 Standard Solution 5, certified reference material, in pyridine (varied)SDS

  • CRM44896ASTM® D6584 1,2,4-Butanetriol Solution, certified reference material, 1000 μg/mL in pyridine, ampule of 5 mLSDS

  • CRM44897ASTM® D6584 Tricaprin Solution, certified reference material, 8000 μg/mL in pyridineSDS

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service