Skip to Content
Merck
All Photos(1)

Key Documents

CRM44897

Supelco

ASTM® D6584 Tricaprin Solution

certified reference material, 8000 μg/mL in pyridine

Synonym(s):

Biodiesel impurity

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€216.00

€216.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
€216.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€216.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

ASTM® D6584

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

kit of 5 mL

concentration

8000 μg/mL in pyridine

application(s)

petroleum

format

single component solution

storage temp.

2-8°C

SMILES string

O(C(COC(=O)CCCCCCCCC)COC(=O)CCCCCCCCC)C(=O)CCCCCCCCC

InChI

1S/C33H62O6/c1-4-7-10-13-16-19-22-25-31(34)37-28-30(39-33(36)27-24-21-18-15-12-9-6-3)29-38-32(35)26-23-20-17-14-11-8-5-2/h30H,4-29H2,1-3H3

InChI key

LADGBHLMCUINGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service