Skip to Content
Merck
All Photos(1)

Key Documents

442847

Supelco

Benzo[a]pyrene-d12

analytical standard

Synonym(s):

3,4-Benzopyrene-d12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20D12
CAS Number:
Molecular Weight:
264.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 10 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

177-180 °C (lit.)

application(s)

environmental

format

neat

mass shift

M+12

storage temp.

2-30°C

SMILES string

[2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c3c([2H])c([2H])c4c([2H])c([2H])c([2H])c5c([2H])c([2H])c2c3c45

InChI

1S/C20H12/c1-2-7-17-15(4-1)12-16-9-8-13-5-3-6-14-10-11-18(17)20(16)19(13)14/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

FMMWHPNWAFZXNH-AQZSQYOVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Badreddine Barhoumi et al.
Marine environmental research, 101, 184-195 (2014-08-12)
In this study, biological responses and contaminant levels in biological tissues were investigated in grass goby fish specimens (Zosterisessor ophiocephalus) collected from five stations in a moderately polluted ecosystem, namely the Bizerte lagoon on the north coast of Tunisia. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service