Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2059

Sigma-Aldrich

Anti-MMP-7 Antibody, clone 2B3 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Matrilysin (UniProt: P09237, also known as EC:3.4.24.23, Matrin, Matrix metalloproteinase-7, MMP-7, Pump-1 protease, Uterine metalloproteinase)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
ICC
WB
clone:
2B3, recombinant monoclonal
species reactivity:
human
citations:
technique(s):
ELISA: suitable
immunocytochemistry: suitable
western blot: suitable
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B3, recombinant monoclonal

description

2B3 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 29.68 kDa
observed mol wt ~25 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... MMP7(4316)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2B3 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects MMP-7. It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of Human MMP-7.

Application

Quality Control Testing

Evaluated by Western Blotting in HT-29 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected MMP-7 in HT-29 cell lysate.

Tested Applications

Enzyme Immunoassay (ELISA) Analysis: A serial of dilutions from a representative lot detected recombinant Human MMP-7.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MMP-7 in HT-29 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Matrilysin (UniProt: P09237; also known as EC:3.4.24.23, Matrin, Matrix metalloproteinase-7, MMP-7, Pump-1 protease, Uterine metalloproteinase) is encoded by the MMP7 (also known as MPSL1, PUMP1) gene (Gene ID:  4316) in human. MMPs are a family zinc-dependent endoproteinases that degrade collagen and other structural proteins and are responsible for much of the turnover of matrix components. Each MMP contains a protease domain with a conserved HExxGHxxGxxHS/T sequence, where three histidine residues form a complex with a catalytic Zn atom.  They contain a generally conserved sequence PRCGVP around the chelating cysteine that is known as the “cysteine switch.” They are produced as zymogens, with a signal sequence and propeptide segment that must be removed during activation. The propeptide domain contains a conserved cysteine, which chelates the active zinc site. MMP-7 is a calcium and zinc-dependent endopeptidase that is synthesized with a signal peptide (aa 1-17) and a propeptide (aa 18-94) that are subsequently cleaved off to produce the active enzyme. It number of substrates including collagen types IV and X, elastin, fibronectin, gelatin, laminin, and proteoglycans. It is the smallest of all the MMPs and lacks the hemopexin-like domain that is common to other members of the family.  MMP-7 is widely expressed, and its elevated levels are observed in cycling endometrium, colorectal cancers and adenomas, hepatocellular carcinomas, pancreatic carcer, and in about 50% of gliomas. MMP-7 play a vital role in the maintenance of innate immunity in organs such as the lungs and intestines where it proteolytically activates anti-bacterial peptides such as pro-defensins. It is also required for mediating proteolytic release of TNF from macrophages. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Liao, H-Y., et al. (2021). Clin. Biochem. 92; 9-18; Ke, B., et al. (2017). Front. Physiol. 8; 21; Burke, B. (2004). Immunobiology. 209(1-2); 51-56).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service