Skip to Content
Merck
All Photos(1)

Documents

720631

Sigma-Aldrich

2-(Trimethylsilyl)-2,3-pentadiene

Synonym(s):

1,3-Dimethyl-1-(trimethylsilyl)allene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16Si
CAS Number:
Molecular Weight:
140.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.444

density

0.746 g/mL at 25 °C

storage temp.

−20°C

SMILES string

CC=C=C(C)[Si](C)(C)C

InChI

1S/C8H16Si/c1-6-7-8(2)9(3,4)5/h6H,1-5H3

InChI key

JGOGDIIEKVYPFO-UHFFFAOYSA-N

Application

Reactant for:
  • Oxidative cyclization with chlorine to give chloro- and acetoxybenziodoxaboroles and a self-assembled tetrameric macrocyclic structure
  • Preparation of allylic alcohols or enone derivatives using boronic acid as catalyst

Catalyst for:
  • Preparation of acylsulfoximines via N-acylation of sulfoximines with aliphatic acids
  • Direct amide bond formation
  • Regioselective preparation of substituted triazole derivatives via dipolar cycloaddition of unsaturated carboxylic acids with azides

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable Liquids

WGK

WGK 3

Flash Point(F)

15.1 °F

Flash Point(C)

-9.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service