Skip to Content
Merck
All Photos(1)

Documents

Z768545

The Belly Dancer® orbital shaker

with 2nd tier platform, AC/DC input 230 V AC, Schuko plug

Synonym(s):

amplification, cell culture growth, gel staining, immunoprecipitation, membrane rinsing, northern hybridization, phage elution, southern hybridization, western blotting

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

feature

0 ‑ 8 ° platform pitch control

manufacturer/tradename

IBI BDRLS0004

parameter

0-100 rpm speed

W × D × H

15 in. × 15 in. × 9 in.

platform W × L

12 in. × 12 in.

Looking for similar products? Visit Product Comparison Guide

General description

Whether your laboratory applications require gentle, continuous agitation or vigorous mixing. The proven IBI Belly Dancer is your instrument of choice. The unique, undulating motion of the Belly Dancer shaker is designed to provide the optimal movement for multiple applications in today′s Life Science Research laboratory.

The smooth combination of vertical and horizontal orbital motion allows for more rapid exchange of reagents in applications such as staining and de-stainign gels and promotes lower background in blotting techniques. The unique 3-dimensional motion and wide range of speeds also provides superior growth in multiple cell culture and tissue culture applications. The easily adjustable speed and platform pitch angle controls provide a wide range of agitation conditions, from extremely gentle to vigorous.

The Belly Dancer is an exceptionally simple liquid agitation system, consisting of an orbiting platform flexibly anchored at four corners ao that the platform motion, when adjusted by the platform pitch control collar, is an extremely effective combination of rocking platform and orbital shaker. One-dimensional shakers do not have the versatility and range of application that result from the variable speed and adjustable platform pitch of the Belly Dancer.

To use this product in the UK please order Z768588 separately

Legal Information

The Belly Dancer is a registered trademark of IBI Scientific Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service