Skip to Content
Merck
All Photos(1)

Key Documents

Z740272

Aluminum heating/cooling block

Holds 48 x 1.5 mL tubes

Synonym(s):

dry block, tube cooling block, tube heating block

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€204.00

€204.00


Estimated to ship on30 March 2025



Select a Size

Change View
1 EA
€204.00

About This Item

UNSPSC Code:
41102404
NACRES:
NB.35

€204.00


Estimated to ship on30 March 2025


material

aluminum

feature

Holds 48 x 1.5 mL tubes

packaging

pkg of 1 ea

manufacturer/tradename

Diversified Biotech CHAM-3000

technique(s)

PCR: suitable

size

156x76x33 mm

Looking for similar products? Visit Product Comparison Guide

General description

Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.

Application

Aluminum heating/cooling block has been used to cool the tubes after centrifugation.[1]

Features and Benefits

  • Solid anodized aluminum, our chambers offer long-lasting durability and reliability.
  • Use with ice pans/buckets for optimal sample cooling.
  • Keep your tubes cool and maintain stable temperatures throughout your sample handling process.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Takahiro Yamasaki et al.
Bio-protocol, 9(5), e3186-e3186 (2019-03-05)
Glycosylation of asparagine residues is widespread in Eukarya, and occurs in virtually all Archaea and some eubacterial species. A membrane-bound enzyme, oligosaccharyltransferase, catalyzes the transfer of an oligosaccharide chain from a sugar donor (lipid-linked oligosaccharide, LLO) to an asparagine residue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service