Skip to Content
Merck
All Photos(2)

Key Documents

HS120729

PCR 96-well Cooler

blue, pink-purple

Sign Into View Organizational & Contract Pricing

Select a Size

2 EACH
€98.30

€98.30


Estimated to ship on11 April 2025



Select a Size

Change View
2 EACH
€98.30

About This Item

UNSPSC Code:
41121800
NACRES:
NB.77

€98.30


Estimated to ship on11 April 2025


material

blue
pink-purple
polycarbonate lid
polypropylene

feature

96-Well, 0.2 mL and 0.5 mL Tubes, and Assay Plates

packaging

pkg of 2 ea

manufacturer/tradename

Heathrow Scientific 120729

L × W × H

110 mm × 148 mm × 48 mm

Looking for similar products? Visit Product Comparison Guide

General description

The PCR Cooler is ideal for thermosensitive applications, PCR, or cell-based assays, as wellas gentle sample thawing and stopping of reactions. The unit also providesa dependable cooling station for pipetting and sample storage. Perfect fortransporting samples and reactions between preparation, thermal cyclingand post-PCR procedure areas.

Features and Benefits

  • Modern patented design cools faster in freezers and stays cold longer on the bench top
  • PCR Cooler maintains a sample temperature of 0 °C (with 2 hours pre-cooling at -20 °C) for over an hour
  • When the temperature ceiling of 6 °C has been exceeded, users are signaled by a contrasting color change of the cooler, or even a single well
  • Cools industry standard format 0.2 mL and 0.5 mL PCR tubes, strips and single format, as well as most 96 well plates, in both standard and low profile well shape
  • Better alternative then ice baths as dry incubation reduces the risk of contamination
  • Freeze cooler in an upright position enabling cold storage of tubes
  • Well design prohibits the freezing of samples
  • Fully sealed and sonic welded cooler keeps coolant contained offering worry free usage
  • Save space with compact footprint and stacking ability
  • The lid is ambidextrous; it can be switched to accommodate left or right-hand orientation and is detachable
  • Robust cooler made from engineered polypropylene and the lid is made from polycarbonate
  • US Patent Pending, Community Design No: 004101756-0001-0002
  • Two Year Warranty

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service