Skip to Content
Merck
All Photos(1)

Key Documents

Z720542

Nalgene® diagnostic bottles, style 2035

capacity 20 mL

Synonym(s):

hdpe bottle, hdpe bottles, nalgene bottle

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
€82.90
100 EA
€307.00

€82.90


Check Cart for Availability


Select a Size

Change View
20 EA
€82.90
100 EA
€307.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

€82.90


Check Cart for Availability

material

high-density polyethylene cap
round transparent PETG bottle

Joint

Joints threaded neck (20-415)

packaging

pack of 20 ea

manufacturer/tradename

Nalgene® 2035-0020

bottle diam. × H

30 mm × 65 mm

capacity

20 mL

Looking for similar products? Visit Product Comparison Guide

General description

Nalgene® diagnostic bottles, style 2035 can be used to perform sterile sampling, storage, and shipment of reagents and buffer solutions. These bottles are made up of non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white high-density polyethylene (HDPE ) closures.

Features and Benefits

  • Sterile to 10-6 SAL
  • Includes a leakproof closure for sample protection.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service