Key Documents
Z720399
Nalgene® centrifuge tubes, Oak Ridge style 3115
nominal capacity 50 mL
Synonym(s):
Nalgene centrifuge tube, plastic centrifuge tube
Select a Size
10 EA
€159.00
100 EA
€1,600.00
€159.00
Estimated to ship on12 May 2025
Select a Size
Change View
10 EA
€159.00
100 EA
€1,600.00
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
€159.00
Estimated to ship on12 May 2025
material
polysulfone
round bottom tube
screw top cap (Linerless)
transparent tube
feature
RCF 50,000 × g
packaging
pack of 10
case of 100
manufacturer/tradename
Nalgene 3115-0050
nominal capacity
50 mL
General description
Strong and tough like polycarbonate but polysulfone much more acid- and base-resistant. Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)