Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
10 ea | Check Cart for Availability | € 120,00 |
100 ea | Check Cart for Availability | € 1.200,00 |
About This Item
€ 120,00
material
cap (linerless closures), conical bottom tube, polypropylene , transparent tube
feature
RCF 50,000 × g
packaging
pack of 10, case of 50
manufacturer/tradename
Nalgene 3148-0050
nominal capacity
35 mL
General description
Features and Benefits
- Silk-screened graduations from 5 to 35 mL in 1 mL increments.
- Translucent polypropylene copolymer tubes offer superior resistance to chemicals.
- Biohazard/Graduated.
- Conical bottoms for enhanced pelleting and separation.
- Able to tolerate forces up to 50,000 xg whether refrigerated or non-refrigerated.
- Leak-proof design.
- Autoclavable numerous times.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| material cap (linerless closures), polypropylene , conical bottom tube, transparent tube | material polysulfone , round bottom tube, screw top cap (Linerless), transparent tube | material polysulfone , round bottom tube, screw top cap (Linerless), transparent tube | material cap (sealing cap), polysulfone , round bottom tube, transparent tube |
| manufacturer/tradename Nalgene 3148-0050 | manufacturer/tradename Nalgene 3115-0030 | manufacturer/tradename Nalgene 3115-0050 | manufacturer/tradename Nalgene 3137-0050 |
| packaging pack of 10, case of 50 | packaging pack of 10, case of 100 | packaging pack of 10, case of 100 | packaging pack of 10, case of 50 |
| feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g | feature RCF 50,000 × g |
| nominal capacity 35 mL | nominal capacity 30 mL | nominal capacity 50 mL | nominal capacity 50 mL |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



