Z674826
Kern ABT analytical balances
weighing capacity 82 or 220 g, resolution: 0.01 or 0.1 mg, linearity: ±0.1 or 0.2 mg
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
Recommended Products
AC/DC input
230 V
material
stainless steel weighing plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
Kern ABT 220-5DNM
availability
not available in USA and Canada
weighing capacity measuring range
82 or 220 g, resolution: 0.01 or 0.1 mg, linearity: ±0.1 or 0.2 mg
plate diam.
80 mm
size
217 mm × 356 mm × 338 mm
Looking for similar products? Visit Product Comparison Guide
General description
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service