Skip to Content
Merck
All Photos(1)

Key Documents

Z660035

Bandelin TICKOPUR R 33 cleaning concentrate

for use in ultrasonic baths

Synonym(s):

detergent

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€25.00
2 L
€38.00
5 L
€76.00

€25.00


Check Cart for Availability


Select a Size

Change View
1 L
€25.00
2 L
€38.00
5 L
€76.00

About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

€25.00


Check Cart for Availability

packaging

pkg of 1 L bottle
pkg of 2 L bottle
pkg of 5 L cannister

manufacturer/tradename

Bandelin 830

General description

Universal cleaner anti-corrosive for laboratory, service and industry, gentle cleaning, mildly alkaline, pH 9.9 (1%), dosage 1 to 5%, 1 to 10 min.

Contamination:

General contamination, oily and greasy residues, soot, ink, drilling-, grinding-, polishing- and lapping residues etc.

Objects to be cleaned:

Glass, ceramics, plastics, rubber, steel, stainless steel, non-ferrous-, precious- and light metals, sieves, pipettes, respirators, PC-boards, glasses. Caution with tin and zinc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service