Skip to Content
Merck
All Photos(1)

Documents

Z660094

Bandelin STAMMOPUR 24 cleaning concentrate

residue-free rinsing, neutral scent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

manufacturer/tradename

Bandelin

availability

available only in EU

General description

Simultaneous intensive cleaning and disinfection Residue-free rinsing, neutral scent. Very gentle to material, anti-corrosive. Free from aldehydes, chlorine and phenols. Extended endurance of the used solution: 3 days. Bactericidal (incl. Tb-B.), fungicidal, virucidal (HBV/HIV). Mildly alkaline, pH 9.4 (1%), dosage 1%, 15 min. (with ultrasound).

Contamination:

General contamination, biofilms, soot, pigments, oil- and fat-containing residues etc.

Objects to be cleaned:

Glass, ceramics, plastics, rubber, steel, stainless steel, non-ferrous-, precious- and light metals, instruments, pipettes, respirators, protective goggles etc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service