Skip to Content
Merck

Skip To

910643

Cleaning solution for Ag ink

Synonym(s):

F47NJET00030 cleaning solution for S-CS01130 Inkjet ink

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 61,60

About This Item

NACRES:
NA.23
UNSPSC Code:
12352103

€ 61,60


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.6 kPa

description

Electronic Solvents

form

liquid

color

colorless

bp

106.1-217.9 °C

Application

This cleaning solution is used to clean product #901083 (Smart’Ink F47NJET00030, Silver nanoparticles ink for inkjet printing).


  • It is a mixture of cyclooctane and alcohols.
  • It is compatible with most substrates including glass and plastic.

Legal Information

Product of Genes′Ink

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
923540901083901090
description

-

description

-

description

-

description

low curing temperature

form

liquid

form

liquid

form

liquid

form

viscous liquid (paste)

vapor pressure

0.6 kPa

vapor pressure

<110 kPa ( 50 °C)

vapor pressure

-

vapor pressure

-

bp

106.1-217.9 °C

bp

101.3-191.1 °C

bp

-

bp

-

color

colorless

color

colorless

color

dark brown to blue

color

-

description

Electronic Solvents

description

Electronic Solvents

description

Electrodes

description

Electrodes


Signal Word

Danger

Hazard Classifications

Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

99.5 °F

Flash Point (°C)

37.5 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

8000-41-7

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters