Key Documents
Z376809
Sigma-Aldrich
CryoTube® work station, autoclavable
pkg of 4
Select a Size
1 PKG
€99.90
€99.90
Check Cart for Availability
Select a Size
Change View
1 PKG
€99.90
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
€99.90
Check Cart for Availability
material
red
packaging
pkg of 4
manufacturer/tradename
Simport T315-3
size
4 in. × 8 in. × 1 in.
General description
An autoclavable rack that holds up to 50 microcentrifuge tubes. Each place is identified with an alpha-numeric index. Unique locking system allows for the tubes to be locked into place while caps may be removed with one hand.
Legal Information
CryoTube is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)