BR114860
BRAND® rack for cryogenic tubes
Holds 50 x cryovials
Synonym(s):
Accessories, Cryogenic Tubes, Rack, Sample Storage, Tubes
Select a Size
€97.20
Select a Size
About This Item
€97.20
Recommended Products
material
blue
polypropylene
feature
Holds 50 x cryovials
packaging
pack of 4 ea
manufacturer/tradename
BRAND 114860
technique(s)
cryopreservation: suitable
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service