Skip to Content
Merck
All Photos(1)

Key Documents

Z374911

PCR multiwell plates

size 384 wells, polypropylene, skirt, non-sterile

Synonym(s):

384 multiwell PCR plate, 384 well PCR microplate, 384 well PCR plate, 384 well microtiter plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
material:
colorless polypropylene
polypropylene
size:
384 wells
well volume:
40 μL
well working volume:
25 μL
sterility:
non-sterile
non-sterile
feature:
skirt

material

colorless polypropylene
polypropylene

sterility

non-sterile
non-sterile

feature

skirt

packaging

case of 50 ea

manufacturer/tradename

Sorenson 39620

technique(s)

PCR: suitable

size

384 wells

well volume

40 μL

well working volume

25 μL

suitability

suitable for (PCR, RT-PCR or DNA purification applications)

application(s)

agriculture

Looking for similar products? Visit Product Comparison Guide

General description

384 well PCR plates are skirted for compatibility with automation systems. Wells have raised rims to ensure contact with sealing film and reduce evaporation. Each well has a capacity of 40 μL and a working volume of 25 μL.
A rigid top plate (included) minimizes plate distortion, assures a dependable fit with the PCR thermal cycler, and allows for a leak-proof seal with micro-mats or cap strips. Each well has a maximum capacity of 200 μL.

Features and Benefits

  • Virgin polypropylene
  • Fully autoclavable
  • Certified DNase- and RNase-free
  • Wells have thin walls for rapid temperature equilibration and reduced cycle time

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Reverse transcription (RT) is the process of converting RNA to cDNA using a reverse transcriptase enzyme and dNTPs.

Hot Start dNTPs block DNA polymerase until heat activation, enhancing PCR specificity.

Perform reverse transcription (RT) using a reverse transcriptase enzyme and dNTPs. Use total RNA or a gene-specific approach so that only the RNA of interest is converted to cDNA.

Standard qPCR protocol with up to four detection probes at specified concentrations, simplifying reaction setup with LuminoCt ReadyMix.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service