Skip to Content
Merck
All Photos(1)

Key Documents

BR781358

BRAND® 384-well PCR plate

full skirt, cut corner A12, H12, white

Synonym(s):

PCR, PCR Plates, Plates

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€375.00

€375.00


Check Cart for Availability


Select a Size

Change View
50 EA
€375.00

About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
material:
polypropylene
white
size:
384 well
sterility:
non-sterile
binding type:
non-treated surface
feature:
skirt
full skirt, cut corner A12, H12

€375.00


Check Cart for Availability

material

polypropylene
white

description

D-Nase/R-Nase, DNA-free, suitable for qPCR

sterility

non-sterile

feature

skirt
full skirt, cut corner A12, H12

packaging

pack of 50 ea

manufacturer/tradename

BRAND 781358

technique(s)

PCR: suitable

size

384 well

working volume

30 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The 40 μL wells can hold sample volumes of 2 μL to 30 μL. This helps to reduce reagents costs and shortens cycle times. The plates can be filled using multichannel pipettes or robotic systems. The ultra thin-walled PCR plate design facilitates constant, rapid and precise heat transfer. The smooth vessel interior minimizes the binding of enzymes and nucleic acid to the walls. The raised lips of the wells protect against cross-contamination, allowing a reliable seal with the sealing mat tailored to the plates.

Features and Benefits

  • For applications in automation and high-throughput analyses
  • Rapid heat transfer through thin vessel walls
  • Can be sealed reliably with self-adhesive sealing films

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service