BR781350
BRAND® 96-well PCR plate
non-skirted, clear, cut corner H12, standard profile
Synonym(s):
PCR, PCR Plates, Plates
Select a Size
€196.00
Select a Size
About This Item
colorless wells
polypropylene
No skirt
blue coding
standard profile
€196.00
Recommended Products
material
clear
colorless wells
polypropylene
description
D-Nase/R-Nase, DNA-free, suitable for qPCR
sterility
non-sterile
feature
skirt: no
No skirt
blue coding
standard profile
packaging
pack of 50 ea
manufacturer/tradename
BRAND 781350
technique(s)
PCR: suitable
size
96 well
working volume
200 μm
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- BRAND® PCR plates consisting of 96 PCR tubes (0.2 mL), with alphanumeric coding.
- Ultra-thin wall design facilitates rapid and steady heat transfer.
- Smooth interior tube surface minimizes binding of enzymes and nucleic acids.
- Raised rims of wells protect against cross-contamination and provide tight seal particularly in combination with BRAND´s new sealing mats.
- Furthermore these sealing mats can be easily pierced with standard pipette tips.
- The PCR-plates can also be sealed with traditional self-adhesive sealing films.
- Skirted plates are available with bar code on request.
- Suitable for all standard thermal cyclers.
- Compatible with cap strips and sealing films to protect against evaporation and contamination.
- Ideal for PCR and qPCR applications.
- Fully-skirted plates, are ideal for automation due to their rigidity.
- Unskirted plates for high flexibility.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service