Skip to Content
Merck
All Photos(1)

Documents

Z374881

PCR microtubes with attached caps

capacity 0.65 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.22

material

clear tube
colorless cap
colorless tube
conical bottom
flat cap
polypropylene
polypropylene cap
polypropylene tube

description

Autoclavable

sterility

non-sterile

feature

RNase and DNase free
graduations: no
Push fit

packaging

pack of 250 tubes
case of 4 packs

technique(s)

PCR: suitable

capacity

0.65 mL

Looking for similar products? Visit Product Comparison Guide

General description

Thin-walled for efficient thermal transfer and shorter cycle times; fits all leading thermal cyclers including PerkinElmer, Biometra, MJ Research, Techne, Grant and Strategene (0.65 mL only). Certified DNase- and RNase-free. All tolerate organic solvent reactions and temperatures from -4 to 121 °C.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

The Extract-N-Amp™ Tissue PCR Kit contains all the reagents needed to rapidly extract and amplify genomic DNA from mouse tails and other animal tissues, buccal swabs, hair shafts, and saliva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service