Skip to Content
Merck
All Photos(1)

Key Documents

EP0030124707

Eppendorf® PCR tubes

Forensic DNA grade, 0.2 mL, colorless, pkg of 500 tubes (10 bags of 50 tubes)

Synonym(s):

Eppendorf® PCR tubes, eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
€93.60

€93.60


Check Cart for Availability


Select a Size

Change View
500 EA
€93.60

About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

€93.60


Check Cart for Availability

material

conical bottom polypropylene

sterility

non-sterile

feature

RNase and DNase free
Forensic DNA grade
cap (hinged lid clip)

packaging

pkg of 500 tubes (10 bags of 50 tubes)

manufacturer/tradename

Eppendorf® 0030124707

technique(s)

PCR: suitable

color

colorless

capacity

0.2 mL

Looking for similar products? Visit Product Comparison Guide

General description

Thin-wall PCR tubes with a contamination shield at the front edge of the lid to prevent accidental contact with the inner surface when the tube is opened.
Matt writing surface on lid

Chem/Phys Resistance

Autoclavable

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service