Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 ea | Check Cart for Availability | € 680,00 |
About This Item
NACRES:
NA.52
UNSPSC Code:
41105501
€ 680,00
Check Cart for Availability
General description
The GenElute 96 Vacuum Manifold is designed to provide the rapid parallel purification of nucleic acids. The manifold is capable of processing up to 96 samples simultaneously and eliminates time consuming centrifugation and decanting steps.
The GenElute Vacuum Manifold is used to support the isolation of genomic DNA, RNA and PCR products which can be used for a variety of molecular biology applications.
The GenElute 96 Vacuum Manifold provides spacer sets to accommodate suitable microtiter plates, deep well blocks (round-well and square-well blocks), or microtube racks for collecting samples.
The GenElute Vacuum Manifold is used to support the isolation of genomic DNA, RNA and PCR products which can be used for a variety of molecular biology applications.
The GenElute 96 Vacuum Manifold provides spacer sets to accommodate suitable microtiter plates, deep well blocks (round-well and square-well blocks), or microtube racks for collecting samples.
The GenElute 96 Vacuum Manifold provides spacer sets to accommodate suitable microtiter plates, deep well blocks (round-well and square-well blocks), or microtube racks for collecting samples.
Application
The GenElute Vacuum Manifold is used to support the isolation of genomic DNA, RNA and PCR products which can be used for a variety of molecular biology applications.
Biochem/physiol Actions
The GenElute 96 Vacuum Manifold is designed to provide the rapid parallel purification of nucleic acids. The manifold is capable of processing up to 96 samples simultaneously and eliminates time consuming centrifugation and decanting steps.
Legal Information
Galacton is a trademark of Tropix, Inc.
GenElute is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Already Own This Product?
Protocols
This protocol is designed to allow for high throughput purification of pure genomic DNA from a variety of cultured cells and tissues using the GenElute 96 Well Tissue Genomic DNA Purification Kit.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| VM01-1EA | 04061837481666 |