Skip to Content
Merck

Skip To

UC213

(R)-(+)-Warfarin

≥97% (HPLC)

Synonym(s):

R-(+)-3-Acetonybenzyl)-4-hydroxycoumarin, R-(+)-4-Hydroxy-3-(3-oxo-1-phenybutyl)-2H-1-benzopyran-2-one

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33
UNSPSC Code:
12161501
PubChem Substance ID:
EC Number:
226-908-9
MDL number:
Assay:
≥97% (HPLC)
Form:
solid


assay

≥97% (HPLC)

form

solid

color

white to pale yellow

mp

≥170 °C

solubility

DMF: soluble, DMSO: soluble, ethanol: soluble

storage temp.

2-8°C

SMILES string

[H][C@@](CC(C)=O)(c1ccccc1)C2=C(O)c3ccccc3OC2=O

InChI

1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21H,11H2,1H3/t15-/m1/s1

InChI key

PJVWKTKQMONHTI-OAHLLOKOSA-N

Gene Information

human ... CYP2C9(1559)

General description

The chiral carbon at position 9 produces two enantiomers, R- and S-warfarin. Commercially available warfarin (Coumadin or Jantoven) is a racemic mixture of S- and R-enantiomers.[1]

Application

(R)-(+)-warfarin has been used to investigate whether enantiomers of warfarin stereoselectively interact with pregnane X nuclear receptor (PXR) to upregulate main drug/xenobiotic-metabolizing enzymes cytochrome P450 3A4 (CYP3A4) and cytochrome P450 2C9 (CYP2C9).[1] It has also been used in HepG2 cells to study protein profiles[2]

Biochem/physiol Actions

CYP1A2/3A4 substrate; anticoagulant.
Warfarin acts as a vitamin K antagonist. It is widely used to treat thromboembolic disorders.[1] (R)-warfarin interacts with pregnane X receptor (PXR) and induces cytochrome P450 3A4 (CYP3A4) and CYP2C9 mRNAs expression in primary human hepatocyte cultures.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Preparation Note

(+)-Warfarin is soluble in DMF, DMSO and ethanol.


signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Repr. 1A - STOT RE 1

target_organs

Blood

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

5543-58-8

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters