Skip to Content
Merck

Skip To

T7443

Trizma® Pre-set crystals

BioPerformance Certified, pH 7.2, average Mw 153.8

Synonym(s):

Tris crystals

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12161700
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

BioPerformance Certified

Quality Segment

form

crystalline powder

mol wt

average Mw 153.8

technique(s)

cell culture | mammalian: suitable

impurities

DNase, RNase, Protease, NICKase, none detected, endotoxin, tested, total aerobic microbial count, tested, ≤0.05 A290, ≤0.5% water (Karl Fischer), ≤5 ppm Heavy metals (as Pb)

pH

7.20 (25 °C, 0.1 M)(lit.), 7.2

useful pH range

7.0-9.0

solubility

water: 0.66 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

Application

Dissolve in water to make a 0.1M solution. No pH adjustment is necessary. Guaranteed accuracy: ±0.1 pH units.

Tris has a significant temperature coefficient, so select the mixture to give the desired final pH at the desired temperature.
From 5 °C to 25 °C, the pH decreases an average of 0.03 pH units per °C.
From 25 °C to 37 °C, the pH decreases an average of 0.025 pH units per °C.

If a different concentration is desired, note that there is a concentration effect on the pH of the solution.
The product is cell culture tested. It is also tested for low endotoxin levels and analyzed for the absence of nucleases and proteases.

Preparation Note

Trizma solutions can be autoclaved.

Other Notes

Certain pH electrodes do not give accurate results with Tris buffers.
This product is a pre-mixed combination of Trizma base and Trizma HCl that can be used to prepare a common pH value for Tris buffer.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T8193T8068T7943
technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

grade

BioPerformance Certified

grade

BioPerformance Certified

grade

BioPerformance Certified

grade

BioPerformance Certified

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

crystalline powder

form

crystalline powder

form

crystalline powder

form

crystalline powder

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

solubility

water: 0.66 g/mL, clear, colorless

solubility

water: 0.66 g/mL, clear, colorless

solubility

water: 0.66 g/mL, clear, colorless

solubility

water: 0.66 g/mL, clear, colorless


Still not finding the right product?

Explore all of our products under Trizma® Pre-set crystals


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
T7443-250G04061837373077
T7443-100G04061837373060

Questions

Reviews

No rating value

Active Filters