Skip to Content
Merck

Skip To

T5705

Terrein

from Aspergillus terreus, ≥98% (HPLC)

Synonym(s):

(+)Terrein, Terrain

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 222,00

About This Item

Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Storage condition:
protect from light

€ 222,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Aspergillus terreus

Quality Segment

assay

≥98% (HPLC)

storage condition

protect from light

solubility

H2O: soluble 1-1.10 mg/mL, clear, colorless, DMSO: soluble

storage temp.

−20°C

SMILES string

OC1C(C(=O)C=C1\C=C\C)O

InChI

1S/C8H10O3/c1-2-3-5-4-6(9)8(11)7(5)10/h2-4,7-8,10-11H,1H3/b3-2+

InChI key

MHOOPNKRBMHHEC-NSCUHMNNSA-N

Biochem/physiol Actions

Terrein is a bioactive fungal metabolite that has plant growth inhibition and antibacterial activities. Terrein reduces melanin levels in a dose-dependent manner as well as tyrosinase protein production.
Terrein is a bioactive fungal metabolite that has plant growth inhibition and antibacterial activities. Terrein reduces melanin levels in a dose-dependent manner as well as tyrosinase protein production. In mammals, there are three melanocyte-specific enzymes: tyrosinase, tyrosinase-related protein-1 (TRP-1), and TRP-2 that are involved in tyrosine conversion into melanin (melanogenesis). Terrein inhibits melanin synthesis by reducing tyrosinase production via extracellular signal-regulated protein kinase (ERK) activation. ERK activation is followed by microphthalmia-associated transcription factor (MITF) down regulation, which is required for tyrosinase expression.

Preparation Note

Terrein is soluble in water at 1-1.10 mg/ml and yields a clear, colorless solution. It is also soluble in DMSO. Hence, the product may be reconstituted in water or DMSO.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T5580SML0459T2577
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

H2O: soluble 1-1.10 mg/mL, clear, colorless, DMSO: soluble

solubility

chloroform: soluble 9.80-10.20 mg/mL, clear, colorless to yellow, DMSO: soluble, acetone: soluble, dichloromethane: soluble, ethanol: soluble, ethyl acetate: soluble, methanol: soluble

solubility

DMSO: soluble, acetone: soluble, ethyl acetate: soluble

solubility

DMSO: 10 mg/mL, clear, H2O: insoluble

storage condition

protect from light

storage condition

protect from light, under inert gas

storage condition

protect from light

storage condition

-

biological source

Aspergillus terreus

biological source

Myxococcus fulvus Mx f85

biological source

Penicillium islandicum

biological source

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters