Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 μg | Check Cart for Availability | € 608,00 |
About This Item
Form:
lyophilized powder
Assay:
≥90% (SDS-PAGE)
Biological source:
human
Recombinant:
expressed in E. coli
Mol wt:
39.7 kDa
€ 608,00
Check Cart for Availability
biological source
human
Quality Segment
recombinant
expressed in E. coli
assay
≥90% (SDS-PAGE)
form
lyophilized powder
mol wt
39.7 kDa
UniProt accession no.
application(s)
cell analysis
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... MAPT(4137)
General description
The gene microtubule associated protein tau (MAPT) is localized on human chromosome 17q21.3. It is expressed in neurons, more specifically in axons.
Biochem/physiol Actions
Isoform of Tau, variant 1N3R, having 3 microtubule binding repeats (R) and one amino terminal insert (N).
Microtubule associated protein tau (MAPT) aids in the assembly and maintenance of microtubule structure. Loss of expression of MAPT results in developmental delay and learning disability. The protein has been associated with pathology of Alzheimer′s disease (AD).
Preparation Note
Lyophilized from MES, pH 6.8, containing NaCl and EGTA. When reconstituted in water to a protein concentration of 1 mg/mL, the resulting buffer will have ~50 mM MES, pH 6.8, 100 mM NaCl, and 0.5 mM EGTA.
1 of 1
This Item | |||
|---|---|---|---|
| biological source human | biological source human | biological source rabbit | biological source rabbit |
| recombinant expressed in E. coli | recombinant - | recombinant - | recombinant - |
| assay ≥90% (SDS-PAGE) | assay >90% (SDS-PAGE) | assay - | assay - |
| mol wt 39.7 kDa | mol wt - | mol wt antigen 78 kDa | mol wt antigen 78 kDa |
| form lyophilized powder | form powder | form buffered aqueous solution | form buffered aqueous solution |
| Gene Information human ... MAPT(4137) | Gene Information - | Gene Information human ... MAPT(4137) | Gene Information human ... MAPT(4137) |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



