Skip to Content
Merck
All Photos(1)

Key Documents

SRP0255

Sigma-Aldrich

eIF4E Active human

N-terminal GST-tag, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

CBP, Eukaryotic translation initiation factor 4E, mRNA cap-binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€651.00

€651.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
100 μG
€651.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€651.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

51 kDa

packaging

pkg of 100 μg

concentration

>0.02 mg/mL

technique(s)

cell based assay: suitable

suitability

suitable for molecular biology

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EIF4E(1977)

General description

EIF4E (eukaryotic translation initiation factor 4E) is a m7GpppN (N is any nucleotide) cap-dependent RNA-binding protein.[1] It is one of the three subunits of the translation initiation factor complex eIF4F, and is the least common of all initiation factors.[2][3]

Application

Suitable substrate for MNK1 kinase.

Biochem/physiol Actions

EIF4E (eukaryotic translation initiation factor 4E) is a part of eIF4F complex, which is responsible for the recruitment of ribosomes to the capped end of mRNAs during mRNA translation. It acts as the rate-limiting element in cap-dependent mRNA translation in eukaryotic cells. It promotes the translation of pro-oncogenic mRNAs, including vascular endothelial growth factor (VEGF), cyclin D3, and Mcl-1 (induced myeloid leukemia cell differentiation). Up-regulation of this protein is linked with various cancers, such as prostate cancer, breast cancer, and lymphoma. Studies show that this protein is overexpressed in melanoma cell lines and is linked with resistance to vemurafenib. Thus, it might have potential as a therapeutic target to suppress melanoma cell proliferation and abrogating vemurafenib resistance.[1][4]

Physical form

Formulated in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, 50% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eukaryotic translation initiation factor 4E (eIF4E) binding site and the middle one-third of eIF4GI constitute the core domain for cap-dependent translation, and the C-terminal one-third functions as a modulatory region.
Morino S, et al.
Molecular and Cellular Biology, 20(2), 468-477 (2000)
Human eukaryotic translation initiation factor 4G (eIF4G) recruits mnk1 to phosphorylate eIF4E.
Pyronnet S, et al.
The Embo Journal, 18(1), 270-279 (1999)
Eukaryotic Translation Initiation Factor 4E Is a Feed-Forward Translational Coactivator of Transforming Growth Factor ? Early Protransforming Events in Breast Epithelial Cells.
Decarlo L, et al.
Molecular and Cellular Biology, 35(15), 2597-2609 (2015)
Yao Zhan et al.
The Journal of investigative dermatology, 135(5), 1368-1376 (2015-01-24)
In eukaryotic cells, the rate-limiting component for cap-dependent mRNA translation is the translation initiation factor eIF4E. eIF4E is overexpressed in a variety of human malignancies, but whether it has a role in melanoma remains obscure. We hypothesized that eIF4E promotes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service