Skip to Content
Merck
All Photos(1)

Key Documents

SML2781

Sigma-Aldrich

NV-5440

≥98% (HPLC)

Synonym(s):

2-[4-[5-[[4-(2-Cyanophenyl)-1-piperazinyl]methyl]-2,4-dimethylbenzoyl]-1-piperazinyl]-benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€85.90
25 MG
€349.00

€85.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€85.90
25 MG
€349.00

About This Item

Empirical Formula (Hill Notation):
C31H36N6O3S
CAS Number:
Molecular Weight:
572.72
UNSPSC Code:
12352200
NACRES:
NA.77

€85.90


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(C=C(C(C)=C1)CN2CCN(CC2)C3=C(C=CC=C3)C#N)C(N4CCN(CC4)C5=CC=CC=C5S(N)(=O)=O)=O

InChI key

BREWSTIKWAQLCH-UHFFFAOYSA-N

Biochem/physiol Actions

NV-5440 is a cell penetrant, potent and selective class I glucose transporters (GLUT1) inhibitor that selectively inhibits mTORC1 pathway in vivo. NV-5440 acts as a potent inhibitor of glycolysis in cells.
cell penetrant, potent and selective GLUT1 inhibitor that selectively inhibits mTORC1 pathway in vivo

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong A Kang et al.
Cell chemical biology, 26(9), 1203-1213 (2019-06-25)
The mechanistic target of rapamycin (mTOR) is a central regulator of cellular metabolic processes. Dysregulation of this kinase complex can result in a variety of human diseases. Rapamycin and its analogs target mTORC1 directly; however, chronic treatment in certain cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service